(3-Chlor-4-fluorphenyl)essigsäure

Lieferant: ABCR
Achtung

Synonyme: 3-Chloro-4-fluorophenylacetic acid

F01882AA
ABCRF01882AAEA 0 EUR
ABCRF01882AA
(3-Chlor-4-fluorphenyl)essigsäure
(3-Chlor-4-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188,59 g/mol
MDL: MFCD01631555
CAS-Nummer: 705-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR