2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid

Lieferant: ABCR

F02651IA.0010
ABCRF02651IA.0010EA 0 EUR
ABCRF02651IA.0010
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
Formel: C₈H₃ClF₂O₃
MDL: MFCD01631474
CAS-Nummer: 143096-86-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR