4-(Difluormethoxy)iodbenzol

Lieferant: ABCR

Synonyme: 1-(Difluoromethoxy)-4-iodobenzene

F02847R.0025
ABCRF02847R.0025EA 0 EUR
ABCRF02847R.0025
4-(Difluormethoxy)iodbenzol
4-(Difluormethoxy)iodbenzol
Formel: C₇H₅F₂IO
Molecular Weight: 270,0153 g/mol
MDL: MFCD00085008
CAS-Nummer: 128140-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR