5-Fluor-1H-indazol

Lieferant: ABCR

F03757H.0005
ABCRF03757H.0005EA 0 EUR
ABCRF03757H.0005
5-Fluor-1H-indazol
5-Fluoro-1H-indazole
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
MDL: MFCD04972877
CAS-Nummer: 348-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR