Nitroniumtetrafluoroborat

Lieferant: ABCR

Synonyme: Nitryl tetrafluoroborate

F05270
ABCRF05270EA 0 EUR
ABCRF05270
Nitroniumtetrafluoroborat
Nitroniumtetrafluoroborat
Formel: BF₄NO₂
Molecular Weight: 132,81 g/mol
Schmelzpunkt: 260 °C
Dichte: 1,26 g/cm³ (20 °C)
MDL: MFCD00011432
CAS-Nummer: 13826-86-3
EINECS: 237-533-5
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR