2-Nitro-4-(trifluormethyl)phenylacetonitril

Lieferant: abcr

Synonyme: 2-(2-Nitro-4-(trifluoromethyl)phenyl)acetonitrile

F05299B.0010
ABCRF05299B.0010EA 0 EUR
ABCRF05299B.0010
2-Nitro-4-(trifluormethyl)phenylacetonitril
2-Nitro-4-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₃N₂O₂
MDL: MFCD00045621
CAS-Nummer: 13544-06-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR