3-Chlor-4-fluorbenzamid

Lieferant: ABCR

Synonyme: 3-Chlor-4-fluorbenzoesäureamid

FO1837B
ABCRFO1837BEA 0 EUR
ABCRFO1837B
3-Chlor-4-fluorbenzamid
3-Chlor-4-fluorbenzamid
Formel: C₇H₅ClFNO
MDL: MFCD00153092
CAS-Nummer: 701-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR