3-Chlor-4-fluorzimtsäure

Lieferant: ABCR

Synonyme: 3-(3-Chlor-4-fluorphenyl)propensäure, 3-Chlor-4-fluorcinnamsäure

FO1864RMC
ABCRFO1864RMCEA 0 EUR
ABCRFO1864RMC
3-Chlor-4-fluorzimtsäure
3-Chlor-4-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200,6 g/mol
MDL: MFCD01631393
CAS-Nummer: 155814-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR