2-Chlor-N-methylacetamid

Lieferant: ABCR

FR-1355.0005
ABCRFR-1355.0005EA 0 EUR
ABCRFR-1355.0005
2-Chlor-N-methylacetamid
2-Chlor-N-methylacetamid
Formel: C₃H₆ClNO
Molecular Weight: 107,54 g/mol
MDL: MFCD00018913
CAS-Nummer: 96-30-0
EINECS: 202-497-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR