3-(2-Hydroxyethyl)pyridin

Lieferant: ABCR

Synonyme: 2-(3-Pyridyl)ethanol

KM09875CB KM09875DA KM09875.0001
MAYBKM09875CBEA 0 EUR
MAYBKM09875CB MAYBKM09875DA ABCRKM09875.0001
3-(2-Hydroxyethyl)pyridin
3-(2-Hydroxyethyl)pyridine
Formel: C₇H₉NO
Molecular Weight: 123,15 g/mol
MDL: MFCD00831047
CAS-Nummer: 6293-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR