1-(3-Aminopropyl)piperidin

Lieferant: abcr

Synonyme: 3-(Piperidin-1-yl)propan-1-amine

MK362.0010
ABCRMK362.0010EA 0 EUR
ABCRMK362.0010
1-(3-Aminopropyl)piperidin
1-(3-Aminopropyl)piperidin
Formel: C₈H₁₈N₂
Molecular Weight: 142.2419 g/mol
MDL: MFCD00023784
CAS-Nummer: 3529-08-6
EINECS: 222-557-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR