1-(4-Pyridyl)piperazin

Lieferant: ABCR

Synonyme: 4-(1-Piperazinyl)pyridin

MK371.0005
ABCRMK371.0005EA 0 EUR
ABCRMK371.0005
1-(4-Pyridyl)piperazin
1-(4-Pyridyl)piperazin
Formel: C₉H₁₃N₃
Molecular Weight: 163,22 g/mol
Siedepunkt: 195…200 °C (0,3 mmHg)
Schmelzpunkt: 140 °C
MDL: MFCD00040745
CAS-Nummer: 1008-91-9
EINECS: 213-764-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR