Methyl-4-cyanbenzoat
Lieferant: abcr
Synonyme:
4-Cyanbenzoesäuremethylester
MK480.0025
ABCRMK480.0025EA
0
EUR
ABCRMK480.0025
Methyl-4-cyanbenzoat
Methyl-4-cyanbenzoat
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol Siedepunkt: 275 Schmelzpunkt: 67 |
MDL:
MFCD00001823 CAS-Nummer: 1129-35-7 EINECS: 214-443-4 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...