4-Amino-1-methylpiperidin

Lieferant: ABCR

Synonyme: 1-Methylpiperidin-4-amine

MK534.0010
ABCRMK534.0010EA 0 EUR
ABCRMK534.0010
4-Amino-1-methylpiperidin
4-Amino-1-methylpiperidin
Formel: C₆H₁₄N₂
Molecular Weight: 114,1888 g/mol
MDL: MFCD01570549
CAS-Nummer: 41838-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR