1-Boc-3-(methylamino)piperidin

Lieferant: ABCR

Synonyme: 3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine

NHC-3123.0001
ABCRNHC-3123.0001EA 0 EUR
ABCRNHC-3123.0001
1-Boc-3-(methylamino)piperidin
1-Boc-3-(methylamino)piperidin
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,31 g/mol
MDL: MFCD04114980
CAS-Nummer: 392331-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR