Bis(triphenylphosphin)palladium(II)chlorid

Lieferant: abcr

PD-4701.0025
ABCRPD-4701.0025EA 0 EUR
ABCRPD-4701.0025
Bis(triphenylphosphin)palladium(II)chlorid
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR