Tri-tert-butylphosphin 10% in Toluol

Lieferant: ABCR

Synonyme: Tris(1,1-dimethylethyl)phosphin

S15-5811TO.50G
ABCRS15-5811TO.50GEA 0 EUR
ABCRS15-5811TO.50G
Tri-tert-butylphosphin 10% in Toluol
Tri-tert-butylphosphin
Formel: C₁₂H₂₇P
Molecular Weight: 202,32 g/mol
MDL: MFCD00015006
CAS-Nummer: 13716-12-6
EINECS: 237-266-4
UN: 2845
ADR: 4.2,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR