Natrium-tert-butanolat ≥98%
Lieferant: ABCR
|
Gefahr
|
Synonyme:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
S931022.0100
ABCRS931022.0100EA
0
EUR
ABCRS931022.0100
Natrium-tert-butanolat ≥98%
Natrium-tert-butanolat
Alkoxide
Formel:
C₄H₉NaO Molecular Weight: 96,1 g/mol |
MDL:
MFCD00040575 CAS-Nummer: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...