1,1,1,3,5,5,5-Heptamethyltrisiloxan

Lieferant: ABCR

Synonyme: 3H-Heptamethyltrisiloxan

SIB1844.0.0010
ABCRSIB1844.0.0010EA 0 EUR
ABCRSIB1844.0.0010
1,1,1,3,5,5,5-Heptamethyltrisiloxan
1,1,1,3,5,5,5-Heptamethyltrisiloxan
Formel: C₇H₂₂O₂Si₃
Molecular Weight: 222,51 g/mol
MDL: MFCD00048000
CAS-Nummer: 1873-88-7
EINECS: 217-496-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR