Dibutylbis(2,4-pentandionato-O,O')zinn

Lieferant: ABCR

Synonyme: Di-n-butyltinbis(acetylacetonate), Di-n-butyltin bis(2,4-pentanedionate), (OC-6-11)-Dibutylbis(2,4-pentanedionato-κO2,κO4)Tin

SND2950.0100
ABCRSND2950.0100EA 0 EUR
ABCRSND2950.0100
Dibutylbis(2,4-pentandionato-O,O')zinn
Dibutylbis(2,4-pentandionato-O,O')zinn
Formel: C₁₈H₃₂O₄Sn
Molecular Weight: 431,15 g/mol
MDL: MFCD00077994
CAS-Nummer: 22673-19-4
EINECS: 245-152-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR