Acroleindiethylacetal ≥95%

Lieferant: ABCR

Synonyme: Acroleinacetal, 3,3-Diethoxypropen

TCA0640.0500
ABCRTCA0640.0500EA 0 EUR
ABCRTCA0640.0500
Acroleindiethylacetal ≥95%
Acroleindiethylacetal
Formel: C₇H₁₄O₂
Molecular Weight: 130,19 g/mol
Siedepunkt: 121°C…125°C
Dichte: 0,850 g/ml
Flammpunkt: 15°C
MDL: MFCD00009239
CAS-Nummer: 3054-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR