cis-4-Aminocyclohexancarbonsäure ≥98,0% (durch GC, titrimetrische Analyse)
Lieferant: ABCR
TCA1448.0005
ABCRTCA1448.0005EA
0
EUR
ABCRTCA1448.0005
cis-4-Aminocyclohexancarbonsäure ≥98,0% (durch GC, titrimetrische Analyse)
cis-4-Aminocyclohexanecarboxylic acid
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,19 g/mol |
MDL:
MFCD00191730 CAS-Nummer: 3685-23-2 |
Spezifikation Testergebnisse
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...