5-Chlortryptamin Hydrochlorid ≥97%

Lieferant: ABCR
Achtung

Synonyme: 5-Chloro-3-(2-aminoethyl)indole Hydrochloride

TCC1145.0005
ABCRTCC1145.0005EA 0 EUR
ABCRTCC1145.0005
5-Chlortryptamin Hydrochlorid ≥97%
5-Chlortryptamin Hydrochlorid
Formel: C₁₀H₁₂Cl₂N₂
Molecular Weight: 231,12 g/mol
Schmelzpunkt: 290°C
MDL: MFCD00051990
CAS-Nummer: 942-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR