trans-1,4-Diaminocyclohexan ≥98%
Lieferant: abcr
|
Gefahr
|
Synonyme:
trans-1,4-Cyclohexandiamin
TCC1426.0025
ABCRTCC1426.0025EA
0
EUR
ABCRTCC1426.0025
trans-1,4-Diaminocyclohexan ≥98%
trans-1,4-Diaminocyclohexane
Formel:
C₆H₁₄N₂ Molecular Weight: 114.19 g/mol Siedepunkt: 197°C Schmelzpunkt: 71°C |
MDL:
MFCD00075174 CAS-Nummer: 2615-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...