2-Chlor-4,6-didimethoxypyrimidin

Lieferant: ABCR

TCC1433
ABCRTCC1433EA 0 EUR
ABCRTCC1433
2-Chlor-4,6-didimethoxypyrimidin
2-Chlor-4,6-didimethoxypyrimidin
Formel: C₆H₇ClN₂O₂
Molecular Weight: 174,58 g/mol
MDL: MFCD00274530
CAS-Nummer: 13223-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR