2,5-Dimethylbenzolsulfonylchlorid

Lieferant: ABCR

Synonyme: p-Xylol-2-sulfonsäurechlorid, p-Xylol-2-sulfonylchlorid, 2,5-Dimethylbenzolsulfochlorid, 2,5-Dimethylbenzol-1-sulfonylchlorid, 2,5-Dimethylbenzolsulfonsäurechlorid

TCD0683
ABCRTCD0683EA 0 EUR
ABCRTCD0683
2,5-Dimethylbenzolsulfonylchlorid
2,5-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Molecular Weight: 204,68 g/mol
MDL: MFCD00024875
CAS-Nummer: 19040-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR