2,3-Dimethoxyanilin

Lieferant: ABCR

Synonyme: (2,3-Dimethoxyphenyl)amine

TCD2284.0005
ABCRTCD2284.0005EA 0 EUR
ABCRTCD2284.0005
2,3-Dimethoxyanilin
2,3-Dimethoxyanilin
Formel: C₈H₁₁NO₂
Molecular Weight: 153,18 g/mol
MDL: MFCD00180769
CAS-Nummer: 6299-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR