Methyl-1-naphthoat

Lieferant: ABCR

Synonyme: 1-Naphthalenecarboxylic acid methyll ester, 1-Naphthoic acid methyll ester, Methyl 1-Naphthalenecarboxylate, Methyl 1-naphthoate

TCN0615.0025
ABCRTCN0615.0025EA 0 EUR
ABCRTCN0615.0025
Methyl-1-naphthoat
Methyl-1-naphthoat
Formel: C₁₂H₁₀O₂
Molecular Weight: 186,2066 g/mol
MDL: MFCD00094243
CAS-Nummer: 2459-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR