4-Acetamidobenzylalkohol

Lieferant: ABCR

Synonyme: N-[4-(Hydroxymethyl)phenyl]acetamide, 4-Acetamidobenzyl alcohol, N-(4-(Hydroxymethyl)phenyl)acetamide

A1052.5 A1052.25 TWA1052.0005
XXXXA1052.5EA 0 EUR
XXXXA1052.5 XXXXA1052.25 ABCRTWA1052.0005
4-Acetamidobenzylalkohol
4-Acetamidobenzylalkohol
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
MDL: MFCD00016868
CAS-Nummer: 16375-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR