3-(2-Chlorphenyl)propionsäure

Lieferant: ABCR

Synonyme: 3-(2-Chlorophenyl)propanoic acid

C2920.5 C2920.25 TWC2920.0005
XXXXC2920.5EA 0 EUR
XXXXC2920.5 XXXXC2920.25 ABCRTWC2920.0005
3-(2-Chlorphenyl)propionsäure
3-(2-Chlorphenyl)propionsäure
Formel: C₉H₉ClO₂
MDL: MFCD00016547
CAS-Nummer: 1643-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR