3-(3-Chlorphenyl)propionsäure

Lieferant: ABCR

C2925.5 C2925.25 TWC2925.0005
XXXXC2925.5EA 0 EUR
XXXXC2925.5 XXXXC2925.25 ABCRTWC2925.0005
3-(3-Chlorphenyl)propionsäure
3-(3-Chlorphenyl)propionsäure
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
MDL: MFCD01310801
CAS-Nummer: 21640-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR