7-Methoxybenzofuran

Lieferant: ABCR

M1262.1 M1262.5 TWM1262.0005
XXXXM1262.1EA 0 EUR
XXXXM1262.1 XXXXM1262.5 ABCRTWM1262.0005
7-Methoxybenzofuran
7-Methoxybenzofuran
Formel: C₉H₈O₂
Molecular Weight: 148,16 g/mol
MDL: MFCD01310825
CAS-Nummer: 7168-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR