Anti-H1L Mouse Monoclonal Antibody

Lieferant: Abgent
AM1012A
ABGEAM1012AEA 354 EUR
ABGEAM1012A
Anti-H1L Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: H1L
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR