Anti-TBB1 Mouse Monoclonal Antibody

Lieferant: Abgent
AM1013A
ABGEAM1013AEA 354 EUR
ABGEAM1013A
Anti-TBB1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TBB1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR