Anti-MUC1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP10097A
ABGEAP10097AEA 354 EUR
ABGEAP10097A
Anti-MUC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human