Anti-FTSJ3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12057C
ABGEAP12057CEA 354 EUR
ABGEAP12057C
Anti-FTSJ3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FTSJ3 (FtsJ homolog 3 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR