Anti-PIAS4 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1249A
ABGEAP1249AEA 354 EUR
ABGEAP1249A
Anti-PIAS4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIAS4 (protein inhibitor of activated STAT, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR