Anti-ZNF222 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP13483A
ABGEAP13483AEA 354 EUR
ABGEAP13483A
Anti-ZNF222 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF222 (zinc finger protein 222)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human