Anti-IBSP Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14114A
ABGEAP14114AEA 354 EUR
ABGEAP14114A
Anti-IBSP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IBSP (integrin-binding sialoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR