Anti-CRTC2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14167B
ABGEAP14167BEA 354 EUR
ABGEAP14167B
Anti-CRTC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CRTC2 (CREB regulated transcription coactivator 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR