Anti-CCDC50 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14241C
ABGEAP14241CEA 354 EUR
ABGEAP14241C
Anti-CCDC50 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC50 (coiled-coil domain containing 50)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR