Anti-DNMT3A Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14999C
ABGEAP14999CEA 354 EUR
ABGEAP14999C
Anti-DNMT3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR