Anti-COX7A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16006C
ABGEAP16006CEA 354 EUR
ABGEAP16006C
Anti-COX7A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COX7A1 (cytochrome c oxidase subunit VIIa polypeptide 1 (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR