Anti-MFAP2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP16274A
ABGEAP16274AEA 354 EUR
ABGEAP16274A
Anti-MFAP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MFAP2 (microfibrillar-associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human