Anti-MS4A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16973B
ABGEAP16973BEA 354 EUR
ABGEAP16973B
Anti-MS4A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR