Anti-SNAI1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP2054D
ABGEAP2054DEA 354 EUR
ABGEAP2054D
Anti-SNAI1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNAI1 (snail family zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human