Anti-ARL3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP2306B
ABGEAP2306BEA 354 EUR
ABGEAP2306B
Anti-ARL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARL3 (ADP-ribosylation factor-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR