Anti-CC50B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP4968A
ABGEAP4968AEA 354 EUR
ABGEAP4968A
Anti-CC50B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CC50B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR