Anti-BCAN Rabbit Polyclonal Antibody

Lieferant: Abgent
AP5915B
ABGEAP5915BEA 354 EUR
ABGEAP5915B
Anti-BCAN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCAN (brevican)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR