Anti-PARP12 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6298B
ABGEAP6298BEA 354 EUR
ABGEAP6298B
Anti-PARP12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PARP12 (poly (ADP-ribose) polymerase family, member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR